2,7-Dichlorofluorene, 97%, Thermo Scientific Chemicals
2,7-Dichlorofluorene, 97%, Thermo Scientific Chemicals
2,7-Dichlorofluorene, 97%, Thermo Scientific Chemicals
2,7-Dichlorofluorene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,7-Dichlorofluorene, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB24717.065 g
Catalog number ALFB24717.06
Price (MYR)
992.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
992.00
EA
Add to cart
Specifications
Chemical Name or Material2,7-Dichlorofluorene
CAS7012-16-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c