2-(Trifluoromethyl)benzaldehyde, 98%
2-(Trifluoromethyl)benzaldehyde, 98%
2-(Trifluoromethyl)benzaldehyde, 98%
2-(Trifluoromethyl)benzaldehyde, 98%
Thermo Scientific Chemicals
2-(Trifluoromethyl)benzaldehyde, 98%
CAS: 447-61-0 | C8H5F3O | 174.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB24591.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-(Trifluoromethyl)benzaldehyde
CAS447-61-0
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.
Solubility
Soluble in most organic solvents.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only
General References:
  1. N Baindur; A Rutledge; D J Triggle. A homologous series of permanently charged 1,4-dihydropyridines: novel probes designed to localize drug binding sites on ion channels. Journal of Medicinal Chemistry. 1993, 36 (23), 3743-3745.
  2. J Alzeer, J Chollet; I Heinze-Krauss; C Hubschwerlen; H Matile; R G Ridley. Phenyl beta-methoxyacrylates: a new antimalarial pharmacophore. Journal of Medicinal Chemistry. 2000, 43 (4), 560-568.