Tetrabromophthalic anhydride, 98%
Tetrabromophthalic anhydride, 98%
Tetrabromophthalic anhydride, 98%
Tetrabromophthalic anhydride, 98%
Thermo Scientific Chemicals
Tetrabromophthalic anhydride, 98%
CAS: 632-79-1 | C8Br4O3 | 463.701 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24550.22100 g
Catalog number ALFB24550.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrabromophthalic anhydride
CAS632-79-1
Health Hazard 1H315-H317-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c