1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFB24492.065 g
Catalog number ALFB24492.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Amino-2-naphthol hydrochloride
CAS1198-27-2
Health Hazard 1H315-H319-H335-H341
Health Hazard 2GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.