Potassium sulfamate, 98%
Potassium sulfamate, 98%
Potassium sulfamate, 98%
Potassium sulfamate, 98%
Thermo Scientific Chemicals
Potassium sulfamate, 98%
CAS: 13823-50-2 | H2KNO3S | 135.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB24447.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium sulfamate
CAS13823-50-2
ColorWhite
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. A. S. Ermakov.; S. A. Serkov.; V. A. Tartakovskii.; T. S. Novikova.; L. I. Khmel'nitskii. Use of potassium sulfamate in the synthesis of heterocyclic nitramines.Chemistry of Heterocyclic Compounds. 1994, 30, (8),976-978 .