1,4-Dimethylpiperazine, 98%
1,4-Dimethylpiperazine, 98%
1,4-Dimethylpiperazine, 98%
1,4-Dimethylpiperazine, 98%
Thermo Scientific Chemicals
1,4-Dimethylpiperazine, 98%
CAS: 106-58-1 | C6H14N2 | 114.192 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24419.22100 g
Catalog number ALFB24419.22
Price (MYR)
521.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
521.00
EA
Add to cart
Specifications
Chemical Name or Material1,4-Dimethylpiperazine
CAS106-58-1
Health Hazard 1H225-H290-H302-H314-H318
Health Hazard 2GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P210-P233-P234-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P372-P374-P380-P501c