4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%
Thermo Scientific Chemicals
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%
CAS: 116-63-2 | C10H9NO4S | 239.245 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24339.1425 g
Catalog number ALFB24339.14
Price (MYR)
651.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
651.00
EA
Add to cart
Specifications
Chemical Name or Material4-Amino-3-hydroxy-1-naphthalenesulfonic acid
CAS116-63-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.
Solubility
Soluble in bases. Insoluble in water.
Notes
Store away from strong oxidizers, strong bases, acid anhydrides, acid chlorides.
RUO – Research Use Only
General References:
  1. Khajavi Mohammad S.; Mohammadi, Ali A. Reaction of 4-amino-3-hydroxy-1-naphthalenesulfonic acid with orthoesters: a new facile one-pot synthesis of 2-substituted naphth[1,2-d]oxazole-5-sulfonates. Journal of Chemical Research. 2002, 2002 (3), 136-138.
  2. T. Kikuchi; T. Furuta and S. Kojima. Membrane localization and demonstration of isoforms of nucleoside triphosphate hydrolase from Toxoplasma gondii. Parasitology. 2001, 122 (1), 15-23.
  3. Key component of the Fiske and Subbarow Reducer for colorimetric determination of phosphate: J. Biol. Chem., 66, 375 (1925); 234, 466 (1959). Reagent for spectrophotometric determination of Si: Microchem. J., 15, 441 (1970).