Zincon monosodium salt
Zincon monosodium salt
Zincon monosodium salt
Zincon monosodium salt
Thermo Scientific Chemicals
Zincon monosodium salt
CAS: 62625-22-3 | C20H16N4NaO6S | 463.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFB24256.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZincon monosodium salt
CAS62625-22-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Reagent for spectrophotometric determination of Zn.
  2. Yan, Y.; Yin, M.; Hong, W.; Xu, Z.; Hu, X. B. Imprinted photonic hydrogel pillar for self-reporting water treatment of heavy metal ions. RSC Adv. 2015, 5 (118), 97604-97610.
  3. Li, M.; Wei, D.; Liu, Q.; Liu, W.; Zheng, J.; Sun, H. Flotation separation of copper-molybdenum sulfides using chitosan as a selective depressant. Miner. Eng. 2015, 83, 217-222.