4-Hydroxybenzamide, 98+%
4-Hydroxybenzamide, 98+%
4-Hydroxybenzamide, 98+%
4-Hydroxybenzamide, 98+%
Thermo Scientific Chemicals
4-Hydroxybenzamide, 98+%
CAS: 619-57-8 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB24222.14
Price (MYR)
827.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
827.00
EA
Add to cart
Specifications
Chemical Name or Material4-Hydroxybenzamide
CAS619-57-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c