tert-Butylcyclohexane, 99+%
tert-Butylcyclohexane, 99+%
tert-Butylcyclohexane, 99+%
tert-Butylcyclohexane, 99+%
Thermo Scientific Chemicals
tert-Butylcyclohexane, 99+%
CAS: 3178-22-1 | C10H20 | 140.27 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24126.22100 g
Catalog number ALFB24126.22
Price (MYR)
1,803.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,803.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butylcyclohexane
CAS3178-22-1
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Wing Tsang. Hydrocarbon decomposition. 6. Thermal decomposition of 3,4-dimethylhexane, 2,2,3-trimethylpentane, tert-butylcyclohexane, and related hydrocarbons. J. Phys. Chem. 1972, 76 (2), 143-156.
  2. Sandra Antúnez.; Eusebio Juaristi. Thermodynamics of the Axial ⇌ Equatorial Conformational Equilibria of tert-Butylcyclohexane and tert-Butyl-Substituted Six-Membered Heterocycles. Theoretical Estimation of Non-Zero Entropy Changes. J. Org. Chem. 1996, 61 (18), 6465-6469.