Check Order Status
(1R)-(-)-Camphorquinone, 98%
Element Search
Structure Search
(1R)-(-)-Camphorquinone, 98%
Have Questions?
Catalog Number
Quantity
ALFB24092.06
5 g
Catalog number
ALFB24092.06
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
(1R)-(-)-Camphorquinone
CAS
10334-26-6
Melting Point
197°C to 202°C
Recommended Storage
Ambient temperatures
TSCA
No
View more