Ethylurea, 98%
Ethylurea, 98%
Ethylurea, 98%
Thermo Scientific Chemicals
Ethylurea, 98%
CAS: 625-52-5 | C3H8N2O | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB24040.36
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylurea
CAS625-52-5
Health Hazard 1H302+H312+H332-H335-H341
Health Hazard 3P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
Melting Point88°C to 94°C
View more
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.
Solubility
Soluble in methanol (25 mg/ml), and water (100 mg/ml).
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Hideshi Suzuki; Hisakazu Sunada. Comparison of Nicotinamide, Ethylurea and Polyethylene Glycol as Carriers for Nifedipine Solid Dispersion Systems. Chemical and Pharmaceutical Bulletin. 1997, 45 (10), 1688-1693.
  2. Michael J. Capaldi and John A. Chapman. The C-terminal extrahelical peptide of type I collagen and its role in fibrillogenesis in vitro: Effects of Ethylurea. Biopolymers. 1984, 23 (2), 313-323.