Hexamethyleneimine, 98+%
Hexamethyleneimine, 98+%
Hexamethyleneimine, 98+%
Hexamethyleneimine, 98+%
Thermo Scientific Chemicals
Hexamethyleneimine, 98+%
CAS: 111-49-9 | C6H13N | 99.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFB24007.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHexamethyleneimine
CAS111-49-9
Health Hazard 1H225-H300-H314-H331
Health Hazard 2GHS H Statement
H225-H300-H314-H318-H312-H332
Highly flammable liquid and vapour.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
RUO – Research Use Only
General References:
  1. Isao Mochida.; Seiichiro Eguchi.; Motohiro Hironaka.; Shin-ichi Nagao.; Kinya Sakanishi.; D. Duayne Whitehurst. The effects of seeding in the synthesis of zeolite MCM-22 in the presence of hexamethyleneimine. Zeolites. 1997, 18 (2-3), 142-151.
  2. Douglas X. West.; Deborah S. Galloway. Transition metal ion complexes of thiosemicarbazones derived from 2-acetylpyridine. Part 3. The 3-hexamethyleneimine derivative. Transition Metal Chemistry. 1988, 13 (6), 410-414.