2-Hydroxy-1-naphthaldehyde, 98%
2-Hydroxy-1-naphthaldehyde, 98%
2-Hydroxy-1-naphthaldehyde, 98%
2-Hydroxy-1-naphthaldehyde, 98%
Thermo Scientific Chemicals
2-Hydroxy-1-naphthaldehyde, 98%
CAS: 708-06-5 | C11H8O2 | 172.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB23951.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Hydroxy-1-naphthaldehyde
CAS708-06-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
2-Hydroxy-1-naphthaldehyde is used in Merrified-type synthesis to deterime free amino acid groups in polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydroxy-1-naphthaldehyde is used in Merrified-type synthesis to deterime free amino acid groups in polymers.
Solubility
Soluble in water (40 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under dry inert gas. Incompatible with bases and oxidizing agents.
RUO – Research Use Only
General References:
  1. T. Dziembowska.; Z. Rozwadowski.; A. Filarowski.; P. E. Hansen. NMR study of proton transfer equilibrium in Schiff bases derived from 2-hydroxy-1-naphthaldehyde and 1-hydroxy-2-acetonaphthone. Deuterium isotope effects on 13C and 15N chemical shifts. Magnetic Resonance in Chemistry. 2001, 39 (1), 67-80.
  2. Siang-Guan Teoh.; Guan-Yeow Yeap.; Ching-Ching Loh.; Lai-Wan Foong.; Soon-Beng Teo.; Hoong-Kun Fun. Inner coordination sphere tin(IV) complexes with some O,N,N-terdentate {N-(2-hydroxybenzaldehyde)-1-amino-2-phenyleneimine and N-(2-hydroxy-1-naphthaldehyde)-1-amino-2-phenyleneimine} and O,N,N,O-quadridentate {N,N'-bis(2-hydroxybenzaldehyde)-1,2-phenylenediimine and N,N'-bis(2-hydroxy-1-naphthaldehyde)-1,2-phenylenediimine} Schiff bases. Polyhedron. 1997, 16 (13), 2213-2221.