6-Methoxy-2-naphthaldehyde, 99%
6-Methoxy-2-naphthaldehyde, 99%
6-Methoxy-2-naphthaldehyde, 99%
6-Methoxy-2-naphthaldehyde, 99%
Thermo Scientific Chemicals
6-Methoxy-2-naphthaldehyde, 99%
CAS: 3453-33-6 | C12H10O2 | 186.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23943.065 g
Catalog number ALFB23943.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material6-Methoxy-2-naphthaldehyde
CAS3453-33-6
Melting Point80°C to 82°C
Recommended StorageAmbient temperatures
EINECS Number222-377-2
View more
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Alexander C. Goudie.; Laramie M. Gaster.; Antony W. Lake.; Carl J. Rose.; Patricia C. Freeman.; Bryn O. Hughes.; David Miller. 4-(6-Methoxy-2-naphthyl)butan-2-one and related analogs, a novel structural class of anti-inflammatory compounds. J. Med. Chem. 1978, 28 (12), 1260-1264.
  2. Jerry P. Jasinsk.; Ray J. Butcher.; Anil N. Mayekar.; H. S. Yathirajan.; B. Narayana. Structures of Three Chalcones Derived from 6-Methoxy-2-naphthaldehyde. Journal of Chemical Crystallography. 2009, 39 (3), 157-162.