Isophthalonitrile, 97+%
Isophthalonitrile, 97+%
Isophthalonitrile, 97+%
Isophthalonitrile, 97+%
Thermo Scientific Chemicals
Isophthalonitrile, 97+%
CAS: 626-17-5 | C8H4N2 | 128.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23900.30250 g
Catalog number ALFB23900.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIsophthalonitrile
CAS626-17-5
Health Hazard 1H302+H332
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c