1,8-Diaminooctane, 98%
1,8-Diaminooctane, 98%
1,8-Diaminooctane, 98%
1,8-Diaminooctane, 98%
Thermo Scientific Chemicals
1,8-Diaminooctane, 98%
CAS: 373-44-4 | C8H22Cl2N2 | 217.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23885.1425 g
Catalog number ALFB23885.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,8-Diaminooctane
CAS373-44-4
Health Hazard 1H302-H314-H317-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
View more
1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water 575g/L.
Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, carbon dioxide, air, water/ moisture.
RUO – Research Use Only
General References:
  1. A Tuel; V Gramlich; Ch Baerlocher. Synthesis, crystal structure and characterization of AP2DAO, a new layered aluminophosphate templated by 1,8-diaminooctane molecules. Microporous and Mesoporous Materials. 2001, 47 (2-3), 217-229.
  2. Meza-Laguna, V.; Basiuk, E. V.; Alvarez-Zauco, E.; Acosta-Najarro, D.; Basiuk, V. A. Cross-Linking of C60 Films with 1,8-Diaminooctane and Further Decoration with Silver Nanoparticles. Journal of Nanoscience and Nanotechnology. 2007, 7 (10), 3563-3571.