Tetraethylene glycol, 99%
Tetraethylene glycol, 99%
Tetraethylene glycol, 99%
Tetraethylene glycol, 99%
Thermo Scientific Chemicals
Tetraethylene glycol, 99%
CAS: 112-60-7 | C8H18O5 | 194.227 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23880.0F2500 mL
Catalog number ALFB23880.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylene glycol
CAS112-60-7
Health Hazard 1H315-H320-H341-H360-H373
Health Hazard 3P201-P202-P262-P264b-P280-P302+P352-P305+P351+P338-P308+P313-P314-P332+P313-P362-P501c
Melting Point-4°C
View more
Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.
Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerin, pyridine, aldehydes, carbon tetrachloride and diethyl ether. Immiscible with oil, benzene, toluene, fat and aliphatic hydrocarbons.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Lee, S.; Alexoff, D. L.; Shea, C.; Kim, D.; Schueller, M.; Fowler, J. S.; Qu, W. Tetraethylene glycol promoted two-step, one-pot rapid synthesis of indole-3-[1-11C]acetic acid. Tetrahedron Lett. 2015, 56 (3), 517-520.
  2. Kim, J. H.; Lee, J. H. Synthesis of spherical bismuth nanoparticles by two-step heating of tetraethylene glycol containing solid bismuth(III) carbonates. Mater. Chem. Phys. 2015, 153, 316-322.