2-Amino-5-nitrobenzophenone, 98%
2-Amino-5-nitrobenzophenone, 98%
2-Amino-5-nitrobenzophenone, 98%
2-Amino-5-nitrobenzophenone, 98%
Thermo Scientific Chemicals
2-Amino-5-nitrobenzophenone, 98%
CAS: 1775-95-7 | C13H10N2O3 | 242.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23773.1425 g
Catalog number ALFB23773.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-nitrobenzophenone
CAS1775-95-7
Health Hazard 1H302
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.
Solubility
Soluble in chloroform, dichloromethane, dimethyl sulfoxide, ethyl acetate and methanol. Soluble in water 3 mg/L (20°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. N. R. Ayyangara, et al. A New And Convenient Synthesis Of 4-Amino-3-Nitrobenzophenone And 2-Amino-5-Nitrobenzophenone And Their N-Alkyl Derivatives.Org. Prep. Proced. Int.,1987,19(2-3), 167-172.
  2. P. J. Cox, et al. 2-Amino-5-nitrobenzophenone and 2'-Fluoro-2-methylamino-5-nitrobenzophenone.Acta Cryst.,1998,C54856-859.