1-Pyrrolidinecarbodithioic acid ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid ammonium salt, 98%
Thermo Scientific Chemicals
1-Pyrrolidinecarbodithioic acid ammonium salt, 98%
CAS: 5108-96-3 | C5H8NS2- | 146.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB23731.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Pyrrolidinecarbodithioic acid ammonium salt
CAS5108-96-3
Health Hazard 1H315-H319-H335-H500
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.
Solubility
Soluble in water.
Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chelating ligand useful for the extraction of many metals from aqueous solution.
  2. Zeng, C.; Kong, J.; Zhou, N.; Cui, R.; Li, X. Room temperature ionic liquids and ammonium pyrrolidine dithiocarbamate synergetically enhanced the determination of zinc by chemical vapor generation coupled with flame atomic absorption spectrometry. Anal. Methods 2013, 5, 1564-1569.
  3. Zeng, C.; Zhou, N.; Luo, J. Synergetic enhancement effect of room temperature ionic liquids andammonium pyrrolidine dithiocarbamate on the chemical vapor generation of iron, cobalt and nickel in environmental and biological samples by atomic absorption spectrometric determination. J. Anal. At. Spectrom. 2012, 27, 120-125.