(3-Mercaptopropyl)trimethoxysilane, 95%
(3-Mercaptopropyl)trimethoxysilane, 95%
(3-Mercaptopropyl)trimethoxysilane, 95%
(3-Mercaptopropyl)trimethoxysilane, 95%
Thermo Scientific Chemicals
(3-Mercaptopropyl)trimethoxysilane, 95%
CAS: 4420-74-0 | C6H16O3SSi | 196.336 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFB23726.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point213°C to 215°C
Chemical Name or Material(3-Mercaptopropyl)trimethoxysilane
Melting Point<-50°C
CAS4420-74-0
Health Hazard 1H302-H317
View more
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.
Solubility
Miscible with methanol, ethanol, isopropanol, acetone, benzene, toluene and xylene.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, alcohols and water. Decomposes in water.
RUO – Research Use Only
General References:
  1. Hau, N. Y.; Chang, Y. H.; Feng, S. P. Kinetics Study of Silver Electrocrystallization on (3-mercaptopropyl)trimethoxysilane-grafted Indium Tin Oxide Plastic Substrate. Electrochim. Acta 2015, 158, 121-128.
  2. Mosa, J.; Durán, A.; Aparicio, M. Sulfonic acid-functionalized hybrid organic-inorganic proton exchange membranes synthesized by sol-gel using 3-mercaptopropyl trimethoxysilane (MPTMS). J. Power Sources 2015, 297, 208-216.