4-Vinylbenzeneboronic acid, 97%
4-Vinylbenzeneboronic acid, 97%
4-Vinylbenzeneboronic acid, 97%
4-Vinylbenzeneboronic acid, 97%
Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 97%
CAS: 2156-04-9 | C8H9BO2 | 147.97 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23709.1425 g
Catalog number ALFB23709.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Vinylbenzeneboronic acid
CAS2156-04-9
Health Hazard 1H302-H312-H315-H319-H332-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.
Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Vancoillie, G.; Pelz, S.; Holder, E.; Hoogenboom, R. Direct nitroxide mediated (co)polymerization of 4-vinylphenylboronic acid as route towards sugar sensors. Polym. Chem. 2012, 3 (7), 1726-1729.
  2. Maji, S.; Vancoillie, G.; Voorhaar, L.; Zhang, Q.; Hoogenboom, R. RAFT Polymerization of 4-Vinylphenylboronic Acid as the Basis for Micellar Sugar Sensors. Macromol. Rapid Commun. 2014, 35 (2), 214-220.