n-Butyl vinyl ether, 98%, stab. with 0.01% KOH
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH
Thermo Scientific Chemicals
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH
CAS: 111-34-2 | C6H12O | 100.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23673.AE100 mL
Catalog number ALFB23673.AE
Price (MYR)
348.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
348.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Butyl vinyl ether
Name Notestabilized with 0.01% KOH
CAS111-34-2
Health Hazard 1H225-H315-H319-H335
Health Hazard 2GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Butyl Vinyl Ether is a reagent used in organic chemical reactions and the preparation of pharmaceutical compounds used in anticancer therapy. It is also used in the synthesis of potent inhibitors for PDE5. It is also used in the synthesis of copolymers and as an acrylic/vinyl acetate resin comonomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water 3 g/L (20°C). Soluble in chloroform and methanol.
Notes
Store away from air and oxidizing agents may form peroxides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Donot distill to dryness. Keep it refrigerated. Check the container pressure periodically to prevent peroxide formation.
RUO – Research Use Only
General References:
  1. Franz Karl Fleischmann, et al. Cycloaddition reaction between tretracyanoethylene and n-butyl vinyl ether: The solvent dependence of the volume of activation.Tetrahedron Lett.,1973,14(39), 3773-3776.
  2. Chang-Min Xing, et al. Surface Photo grafting Polymerization of Binary Monomers Maleic Anhydride and n-Butyl Vinyl Ether on Polypropylene Film I. Effects of Principal Factors.Polym. J.,2002,34801-808.