Cobalt(II) thiocyanate, 98+%
Cobalt(II) thiocyanate, 98+%
Cobalt(II) thiocyanate, 98+%
Cobalt(II) thiocyanate, 98+%
Thermo Scientific Chemicals
Cobalt(II) thiocyanate, 98+%
CAS: 3017-60-5 | C2H2CoN2S2 | 177.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALFB23460.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II) thiocyanate
CAS3017-60-5
Health Hazard 1H302+H312+H332-H350
Health Hazard 2GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
View more
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.
Solubility
Soluble in water.
Notes
Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Palion-Gazda, J.; Machura, B.; Lloret, F.; Julve, M. Ferromagnetic Coupling Through the End-to-End Thiocyanate Bridge in Cobalt(II) and Nickel(II) Chains. Crys. Growth Des. 2015, 15 (5), 2380-2388.
  2. Kimer, J. T.; Elliott, C. M. Are High-Potential Cobalt Tris(bipyridyl) Complexes Sufficiently Stable to Be Efficient Mediators in Dye-Sensitized Solar Cells? Synthesis, Characterization, and Stability Tests. J. Phys. Chem. C. 2015, 119 (31), 17502-17514.