2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
Thermo Scientific Chemicals
2,4-Dibromophenol, 98%
CAS: 615-58-7 | C6H4Br2O | 251.905 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23394.22100 g
Catalog number ALFB23394.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dibromophenol
CAS615-58-7
Health Hazard 1H300-H315-H319-H335
Health Hazard 2GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c