Light Green SF Yellowish
Light Green SF Yellowish
Light Green SF Yellowish
Light Green SF Yellowish
Thermo Scientific Chemicals
Light Green SF Yellowish
CAS: 5141-20-8 | C37H34N2Na2O9S3 | 792.844 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23330.1850 g
Catalog number ALFB23330.18
Price (MYR)
1,150.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,150.00
EA
Add to cart
Specifications
Chemical Name or MaterialLight Green SF Yellowish
Melting Point∼288°C (decomposition)
CAS5141-20-8
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H351
Suspected of causing cancer.
View more
Useful as a general plasma stain, as a histological stain, and a cytological counterstain