Specifications
Chemical Name or Material4-Allylanisole
CAS140-67-0
Health Hazard 1H227-H302-H315-H317-H319-H341-H351
Health Hazard 2GHS H Statement
H301-H341-H351-H227-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Suspected of causing cancer.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P210-P261-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P370+P378q-P501c
View more