beta-Propiolactone, 97%
beta-Propiolactone, 97%
beta-Propiolactone, 97%
beta-Propiolactone, 97%
Thermo Scientific Chemicals
beta-Propiolactone, 97%
CAS: 57-57-8 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFB23197.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialbeta-Propiolactone
CAS57-57-8
Health Hazard 1H227-H315-H319-H330-H350
Health Hazard 2GHS H Statement
H330-H350-H227-H315-H319
Health Hazard 3P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P284-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P378q-P501c
View more
Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.
Solubility
Miscible with water and organic solvents.
Notes
Flammable material. Should be stored in a separate safety storage cabinet or room. Keep away from heat and sources of ignition.
RUO – Research Use Only
General References:
  1. Goodman, C. H.; Russell, B. J.; Velez, J. O.; Laven, J. J.; Bagarozzi, Jr. D. A.; Moon, J. L.; Bedi, K.; Johnson, B. W. Production of a Sindbis/Eastern Equine Encephalitis chimeric virus inactivated cell culture antigen. J. Virol. Methods. 2015, 223, 19-24.
  2. Pawar, S.D.; Murtadak, V. B.; Kale, S. D.; Shinde, P. V.; Parkhi, S. S. Evaluation of different inactivation methods for high and low pathogenic avian influenza viruses in egg-fluids for antigen preparation. J. Virol. Methods. 2015, 222, 28-33.