2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
Thermo Scientific Chemicals
2-(Methylamino)ethanol, 99%
CAS: 109-83-1 | C3H9NO | 75.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23180.AP500 mL
Catalog number ALFB23180.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material2-(Methylamino)ethanol
CAS109-83-1
Health Hazard 1H227-H302+H312-H314-H335-H373
Health Hazard 2GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.
Solubility
Miscible with water, alcohol and ether.
Notes
Incompatible with oxidizing agents, acids, copper, copper alloys, zinc and galvanized iron.
RUO – Research Use Only
General References:
  1. Khusnutdinova, J. R.; Garg, J. A.; Milstein, D. Combining Low-Pressure CO2 Capture and Hydrogenation To Form Methanol. ACS Catal. 2015, 5 (4), 2416-2422.
  2. Fytianos, G.; Callot, R.; Svendsen, H. F.; Knuutila, H. K. Quantitative determination of amines used in post-combustion CO2 capture process by ion chromatography. Int. J. Greenhouse Gas Control 2015, 42, 372-378.