3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
Thermo Scientific Chemicals
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
CAS: 3069-29-2 | C8H22N2O2Si | 206.36 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23159.22100 g
Catalog number ALFB23159.22
Price (MYR)
409.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
409.00
EA
Add to cart
Specifications
Boiling Point265°C
Chemical Name or Material3-(2-Aminoethylamino)propylmethyldimethoxysilane
CAS3069-29-2
Health Hazard 1H227-H302-H315-H317-H318
Health Hazard 2GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.