1,5-Diaminopentane, 98%
1,5-Diaminopentane, 98%
1,5-Diaminopentane, 98%
1,5-Diaminopentane, 98%
Thermo Scientific Chemicals
1,5-Diaminopentane, 98%
CAS: 462-94-2 | C5H14N2 | 102.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23039.065 g
Catalog number ALFB23039.06
Price (MYR)
1,014.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,014.00
EA
Add to cart
Specifications
Chemical Name or Material1, 5-Diaminopentane
CAS462-94-2
Health Hazard 1H227-H302+H312-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H227
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
1,5-Diaminopentane is mainly used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Diaminopentane is mainly used in organic synthesis.
Solubility
Soluble in water.
Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
RUO – Research Use Only
General References:
  1. Biljana R Cvetković; Lato L Pezo; Tatjana Tasić; Ljubiša Šarić; Žarko Kevrešan; Jasna Mastilović. The optimisation of traditional fermentation process of white cabbage (in relation to biogenic amines and polyamines content and microbiological profile). Food Chemistry. 2015, 168, 471-477.
  2. Takashi Mimitsuka; Hideki Sawai; Masahiro Hatsu; Katsushige Yamada. Metabolic engineering of Corynebacterium glutamicum for cadaverine fermentation. Bioscience, Biotechnology, and Biochemistry. 2007, 71 (9), 2130-2135.