1,2,4-Benzenetricarboxylic anhydride, 97%
1,2,4-Benzenetricarboxylic anhydride, 97%
1,2,4-Benzenetricarboxylic anhydride, 97%
1,2,4-Benzenetricarboxylic anhydride, 97%
Thermo Scientific Chemicals
1,2,4-Benzenetricarboxylic anhydride, 97%
CAS: 552-30-7 | C9H4O5 | 192.126 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23003.A11 kg
Catalog number ALFB23003.A1
Price (MYR)
566.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
566.00
EA
Add to cart
Specifications
Chemical Name or Material1,2,4-Benzenetricarboxylic anhydride
CAS552-30-7
Health Hazard 1H312-H317-H318-H332-H334-H335
Health Hazard 2GHS H Statement
H334-H318-H335-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P342+P311-P363-P501c
View more
1,2,4-Benzenetricarboxylic anhydride is used to produce good heat-proof, weather-proof and solvent-proof trimellitate plasticizers, to synthesize polyester resins, curing agent of epoxy resins, heat-proof insulated laminas, heat-proof varnishes, stabilizers, dyestuffs, pigments, surfactants. TMA can also be used as accelerant for vulcanization, treating agent of fiber and flocculant of film. It has wide application to high class aerooil, impregnating oil for power capacitors, adhesives for granules, instant adhesives, sizing agents, antismoke agents, dyestuffs, printing inks, medicines and farm chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,4-Benzenetricarboxylic anhydride is used to produce good heat-proof, weather-proof and solvent-proof trimellitate plasticizers, to synthesize polyester resins, curing agent of epoxy resins, heat-proof insulated laminas, heat-proof varnishes, stabilizers, dyestuffs, pigments, surfactants. TMA can also be used as accelerant for vulcanization, treating agent of fiber and flocculant of film. It has wide application to high class aerooil, impregnating oil for power capacitors, adhesives for granules, instant adhesives, sizing agents, antismoke agents, dyestuffs, printing inks, medicines and farm chemicals.
Solubility
Hydrolyzes in water. Soluble in ethanol, acetone, and most other organic solvents.
Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, bases, water/ moisture.
RUO – Research Use Only
General References:
  1. V. Arjunan; Arushma Raja, S. Subramanian; S. Mohan. Vibrational, electronic and quantum chemical studies of 1,2,4-benzenetricarboxylic-1,2-anhydride. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2013, 110 141-150.
  2. Ling Li; Lei Feng; Qinbo Wang and Xi Li. Solubility of 1,2,4-Benzenetricarboxylic Acid in Acetic Acid + Water Solvent Mixtures. J. Chem. Eng. Data. 2008, 53, (1), 298-300.