3-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific™
3-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific™
3-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific™
3-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific™
3-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFB22975.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Chloro-2-fluorobenzaldehyde
CAS85070-48-0
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.