tert-Butyl ethyl ether, 99%
tert-Butyl ethyl ether, 99%
tert-Butyl ethyl ether, 99%
tert-Butyl ethyl ether, 99%
Thermo Scientific Chemicals
tert-Butyl ethyl ether, 99%
CAS: 637-92-3 | C6H14O | 102.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22945.22100 g
Catalog number ALFB22945.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialtert-Butyl ethyl ether
CAS637-92-3
Health Hazard 1H225-H336
Health Hazard 2GHS H Statement
H225-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
View more
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.
RUO – Research Use Only
General References:
  1. Iuga, C.; Osnaya-Soto, L.; Ortiz, E.; Vivier-Bunge, A. Atmospheric oxidation of methyl and ethyl tert-butyl ethers initiated by hydroxyl radicals. A quantum chemistry study. Fuel 2015, 159, 269-279.
  2. Liu, X.; Ito, S.; Wada, Y. Oxidation characteristic and products of ETBE (ethyl tert-butyl ether). Energy 2015, 82, 184-192.