Methoxyacetic acid, 97%
Methoxyacetic acid, 97%
Methoxyacetic acid, 97%
Methoxyacetic acid, 97%
Thermo Scientific Chemicals
Methoxyacetic acid, 97%
CAS: 625-45-6 | C3H6O3 | 90.078 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22917.1850 g
Catalog number ALFB22917.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethoxyacetic acid
CAS625-45-6
Health Hazard 1H302-H314-H335-H360FD
Health Hazard 2GHS H Statement
H360-H314-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Reacts with thionyl chloride on prolonged refluxing, to give chloromethyl methyl ether, reportedly free from the highly carcinogenic bis(chloromethyl)ether: Synthesis, 727 (1984).
  2. świechowicz, B. S.; Szymczak, W.; Budziszewska, B.; Starek, A. Testicular effect of a mixture of 2-methoxyethanol and 2-ethoxyethanol in rats. Pharmacol. Rep. 2015, 67 (2), 289-293.
  3. Grala, A.; Pietkiewicz, M. W.; Wojewódzka, A.; Dabergut, M.; Justyniak, I.; Lewiński, J. Structural Diversity of Ethylzinc Carboxylates. Organometallics 2015, 34 (20), 4959-4964.