(+/-)-2-Aminoheptane, 98+%
(+/-)-2-Aminoheptane, 98+%
(+/-)-2-Aminoheptane, 98+%
(+/-)-2-Aminoheptane, 98+%
Thermo Scientific Chemicals
(+/-)-2-Aminoheptane, 98+%
CAS: 123-82-0 | C7H17N | 115.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22863.22100 g
Catalog number ALFB22863.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-2-Aminoheptane
CAS123-82-0
Health Hazard 1H226-H302+H312+H332-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water ( 9 g/L at 20°C).
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only
General References:
  1. P.Macaudiere; A.Tambute; M.Caude; R.Rosset; M.A.Alembik; I.W.Wainr. Resolution of enentiomeric amides on a pirklet-type chiral stationary phase : A comparison of subcritical fluid and liquid chromatographic approaches. Journal of Chromatography A. 1986, 371,177-193.
  2. J.A.Corbin; Lockhart Burgess Rogers. Improved gas chromatographic separations of enantiomeric secondary amine derivatives. Anal. Chem. 1970, 42,(9), 974-979.