4-Chlorobenzyl bromide, 98+%
4-Chlorobenzyl bromide, 98+%
4-Chlorobenzyl bromide, 98+%
4-Chlorobenzyl bromide, 98+%
Thermo Scientific Chemicals
4-Chlorobenzyl bromide, 98+%
CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22858.065 g
Catalog number ALFB22858.06
Price (MYR)
470.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
470.00
EA
Add to cart
Specifications
Chemical Name or Material4-Chlorobenzyl bromide
CAS622-95-7
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
RUO – Research Use Only
General References:
  1. Masaru Ogata; Hiroshi Matsumoto; Sumio Shimizu; Shiro Kida; Motoo Shiro; Katsuya Tawara. Synthesis and antifungal activity of new 1-vinylimidazoles. J. Med. Chem. 1987, 30,(8), 1348-1354.
  2. Christian Beaulieu; Zhaoyin Wang; Danielle Denis; Gillian Greig; Sonia Lamontagne; Gary O'Neill; Deborah Slipetz; Jennifer Wang. Benzimidazoles as new potent and selective DP antagonists for the treatment of allergic rhinitis. Bioorganic & Medicinal Chemistry Letters. 2004, 14,(12), 3195-3199.