Flavanone, 98+%
Flavanone, 98+%
Flavanone, 98+%
Thermo Scientific Chemicals
Flavanone, 98+%
CAS: 487-26-3 | C15H12O2 | 224.26 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22587.0910 g
Catalog number ALFB22587.09
Price (MYR)
322.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
322.00
EA
Add to cart
Specifications
Chemical Name or MaterialFlavanone
CAS487-26-3
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Melting Point75°C to 79°C