Strontium bromide hexahydrate, 99%
Strontium bromide hexahydrate, 99%
Strontium bromide hexahydrate, 99%
Strontium bromide hexahydrate, 99%
Thermo Scientific Chemicals
Strontium bromide hexahydrate, 99%
CAS: 7789-53-9 | Br2H12O6Sr | 355.518 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22553.22100 g
Catalog number ALFB22553.22
Price (MYR)
537.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
537.00
EA
Add to cart
Specifications
Chemical Name or MaterialStrontium bromide hexahydrate
CAS7789-53-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Strontium bromide hexahydrate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Strontium bromide hexahydrate is used as pharmaceutical intermediate.
Solubility
Soluble in water (2850 g/L at 20°C).
Notes
m.p. 88° (rapidly heated) , 180° -6H{2}O
RUO – Research Use Only
General References:
  1. E.Buzagh-Gere; J.Simon; S.Gal. Characterization of differently bound water by means of the constants of thermal decomposition kinetics. Fresenius' Zeitschrift für analytische Chemie. 1973, 264,(5), 392-396.
  2. W.D Shuster; S.Subler; E.L.McCoy. The influence of earthworm community structure on the distribution and movement of solutes in a chisel-tilled soil. Applied Soil Ecology. 2002, 21,(2), 159-167.