4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB22530.0910 g
Catalog number ALFB22530.09
Price (MYR)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone
CAS4551-69-3
Melting Point90°C to 92°C
Recommended StorageAmbient temperatures
EINECS Number224-918-8