(3-Chloropropyl)trimethoxysilane, 97%
(3-Chloropropyl)trimethoxysilane, 97%
(3-Chloropropyl)trimethoxysilane, 97%
(3-Chloropropyl)trimethoxysilane, 97%
Thermo Scientific Chemicals
(3-Chloropropyl)trimethoxysilane, 97%
CAS: 2530-87-2 | C6H15ClO3Si | 198.718 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB22517.36
Price (MYR)
1,024.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,024.00
EA
Add to cart
Specifications
Boiling Point195°C to 197°C
Chemical Name or Material(3-Chloropropyl)trimethoxysilane
Melting Point-50°C
CAS2530-87-2
Health Hazard 1H227
View more
(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only
General References:
  1. Farook Adam; Hasnah Osman; Kasim Mohammed Hello. The immobilization of 3-(chloropropyl)triethoxysilane onto silica by a simple one-pot synthesis. Journal of Colloid and Interface Science. 2009, 331 (1), 143-147.
  2. M. Čapka. Selective Phosphination of (3-Chloropropyl)triethoxysilane. A New Route to Alkoxysilyl-Substituted Phosphines. Synthesis and Reactivity in Inorganic and Metal-Organic Chemistry. 1977, 7 (4), 347-354.