4-Cyanophenyl 4-n-hexylbenzoate, 99%
4-Cyanophenyl 4-n-hexylbenzoate, 99%
4-Cyanophenyl 4-n-hexylbenzoate, 99%
4-Cyanophenyl 4-n-hexylbenzoate, 99%
Thermo Scientific Chemicals
4-Cyanophenyl 4-n-hexylbenzoate, 99%
CAS: 50793-85-6 | C20H21NO2 | 307.39 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22471.031 g
Catalog number ALFB22471.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Cyanophenyl 4-n-hexylbenzoate
CAS50793-85-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c