4-(Hydroxymethyl)benzoic acid, 98+%
4-(Hydroxymethyl)benzoic acid, 98+%
4-(Hydroxymethyl)benzoic acid, 98+%
4-(Hydroxymethyl)benzoic acid, 98+%
Thermo Scientific Chemicals
4-(Hydroxymethyl)benzoic acid, 98+%
CAS: 3006-96-0 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22183.031 g
Catalog number ALFB22183.03
Price (MYR)
557.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
557.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Hydroxymethyl)benzoic acid
CAS3006-96-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c