2'-Hydroxy-4'-methoxyacetophenone, 99%
2'-Hydroxy-4'-methoxyacetophenone, 99%
2'-Hydroxy-4'-methoxyacetophenone, 99%
2'-Hydroxy-4'-methoxyacetophenone, 99%
Thermo Scientific Chemicals
2'-Hydroxy-4'-methoxyacetophenone, 99%
CAS: 552-41-0 | C9H10O3 | 166.176 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22173.065 g
Catalog number ALFB22173.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2'-Hydroxy-4'-methoxyacetophenone
CAS552-41-0
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c