Polyethylene glycol 1,000
Polyethylene glycol 1,000
Polyethylene glycol 1,000
Polyethylene glycol 1,000
Thermo Scientific Chemicals
Polyethylene glycol 1,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFB22134.30
Price (MYR)
248.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
248.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolyethylene glycol 1000
CAS25322-68-3
Physical FormWaxy Solid
Health Hazard 1H227-H303-H315-H320-H333-H335
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
View more
Polyethylene glycol is used as an excipent in pharmaceutical products. It is used in the precipitation of proteins as well as in the separation and purification of biomolecules and in the induction of cell hybridization. It acts as a fusing agent to enhance the effect of macrophages on hybridoma; as a vascular agent in preclinical work; as an anti-foaming agent in food and as the gate insulator in an electric double-layer transistor to enhance superconductivity in an insulator.