Ficoll∣r 400
Ficoll∣r 400
Ficoll∣r 400
Thermo Scientific Chemicals
Ficoll∣r 400
CAS: 26873-85-8 | C15H27ClO12 | 434.819 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22095.1850 g
Catalog number ALFB22095.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFicoll 400
CAS26873-85-8
Recommended StorageAmbient temperatures
EINECS Number200-334-9
TSCAYes
View more
Ficoll™ 400 is used for cell separation and organelle isolation. It is also used to separate blood to its components like erythrocytes and leukocytes. It acts as a mononuclear cell separator and used in culture media, nucleic acidhybridization, electrophoresis and in immunological studies.