Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Thermo Scientific Chemicals
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
CAS: 6148-75-0 | C9H11NaO3S | 222.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22029.22100 g
Catalog number ALFB22029.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMesitylenesulfonic acid sodium salt hemihydrate
CAS6148-75-0
Melting Point>300°C
Recommended StorageAmbient temperatures
EINECS Number228-157-2