7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
Thermo Scientific Chemicals
7-Diethylamino-4-methylcoumarin, 99%
CAS: 91-44-1 | C14H17NO2 | 231.295 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21938.36500 g
Catalog number ALFB21938.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material7-Diethylamino-4-methylcoumarin
CAS91-44-1
Health Hazard 1H500
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.
Solubility
Soluble in water, aqueous acid solutions, resins, varnishes, vinyls, ether, acetone, ordinary organic solvents, and alcohol.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. T. Gustavsson; L. Cassara; S. Marguet; G. Gurzadyan; P. van der Meulen; S. Pommeret and J.-C. Mialocq. Rotational diffusion of the 7-diethylamino-4-methylcoumarin C1 dye molecule in polar protic and aprotic solvents. Photochem. Photobiol. Sci. 2003, 2, 329-341.
  2. R.W. Yip; Y.X. Wen. Photophysics of 7-diethylamino-4-methylcoumarin: picosecond time-resolved absorption and amplified emission study. Journal of Photochemistry and Photobiology A: Chemistry. 1990, 54 (2), 263-270.