2-Aminobenzenesulfonamide, 98%
2-Aminobenzenesulfonamide, 98%
2-Aminobenzenesulfonamide, 98%
2-Aminobenzenesulfonamide, 98%
Thermo Scientific Chemicals
2-Aminobenzenesulfonamide, 98%
CAS: 3306-62-5 | C6H8N2O2S | 172.202 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21912.065 g
Catalog number ALFB21912.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Aminobenzenesulfonamide
CAS3306-62-5
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point153°C to 157°C