4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™
4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™
4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™
4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™
4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFB21829.1425 g
Catalog number ALFB21829.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminonaphthalene-1-sulfonic acid
CAS84-86-6
Health Hazard 1H302-H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Amino-1-naphthalenesulfonic acid has potent therapeutic applications. Its sodium salt has been used as non-toxic hemostatic. It is also a metabolite of azo dyes.
Solubility
Sparingly soluble in dichloromethane. Soluble in aqueous base.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
General References:
  1. Alonso MC; and Barceló D. Tracing polar benzene-and naphthalenesulfonates in untreated industrial effluents and water treatment works by ion-pair chromatography-fluorescence and electrospray-mass spectrometry. Anal. Chim. Acta., 1999, 400 (1), 211-231.
  2. I Lindh; W J Griffiths; T Bergman; J Sjövall. Charge-remote fragmentation of peptides derivatized with 4-aminonaphthalenesulphonic acid. Rapid Communications in Mass Spectrometry, 1994, 8 (10), 797-803.